missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Adapter Support

Product Code. p-7164752 Shop All Brand Products
Change view
Click to view available options
Color:
Dark Blue
Green
Magenta
Unit Size:
Each
Product Code. Color unitSize
10235032 Magenta Each
10113402 Green Each
10225032 Dark Blue Each
3 options
This item is not returnable. View return policy

Product Code. 10235032

Brand: BRAND™ 26653

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Polypropylene Adapter Support designed for use with accu-jet™ pro.

  • Constructed of polypropylene
  • For use with accu-jet™
TRUSTED_SUSTAINABILITY

Specifications

Color Magenta
Material Polypropylene
Type Adapter Support
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.