missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Polypropylene 96-well F-Bottom Microplates
GREENER_CHOICE

Product Code. 10093241
Change view
Click to view available options
Color:
Black
Clear
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility unitSize
10093241 Clear Sterile Pack of 100
10131992 Clear Non-sterile Pack of 100
10706172 Black Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10093241 Supplier Greiner Bio-One Supplier No. 655261
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from polypropylene for high resistance to common chemicals and thermal stability. Greiner Bio-One–Polypropylene 96 Well Microplates– are ideal for storage of active reagents, patient samples or biomolecules.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Traceability to production through a defined LOT number system
  • Applications: - Long-term storage of active agents - Storage of patient samples in diagnostics - Storage of DNA or RNA, stock cultures
  • Uniform external dimensions
  • Well-to-well spacing 9 mm
  • Alphanumeric well coding High chemical resistance
  • High temperature resistance (-196 to +121°C)
  • Sealable with adhesive films and heat sealer or CapMats
  • Solid bottom
  • Well profile: F-Bottom/chimney well (flat bottom)
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polypropylene
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) Long-term storage of active agents
Quantity 100/Cs.
Temperature Range (Metric) –196 to 121°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.