missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropelene Screw Caps with Lip Seal

Catalog Number 11537033
Click to view available options
Color:
Green
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy
Closure Type Screw Cap
For Use With (Equipment) For reagent and media bottles
Color Green
Description Screw cap with lip seal
Material Polypropylene
Length (Metric) 92.5mm
Quantity 10/PK
Thread Size (Metric) 45mm
Certifications/Compliance ISO R1115 and DIN 168
Autoclavable Autoclavable
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'