missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Polyplast Temp DIN Electrode

Product Code. 10649715
Click to view available options
Length (Metric):
120 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10649715

Brand: Hamilton™ 238404

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Accuracy (mV) 0 ± 20mV
Accuracy (Temperature) 0°C to 60°C
Diameter (Metric) 12 mm
Electrical Requirements 0 ± 20 mV, 57 to 59 mV
Length (Metric) 120 mm
pH Range 0 to 14
Temperature (Metric) Range 0°C to 60°C
Certifications/Compliance Yes, with measured values in Buffer pH 4.01 and pH 7.00
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.