missing translation for 'onlineSavingsMsg'
Learn More

Polyphosphoric Acid, Reagent grade, 115% H3PO4 basis, Honeywell™

Product Code. 15621830 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15621830 1 kg Poly Bottle 1kg
15631830 25 g Poly Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15621830 Supplier Honeywell Research Chemicals Supplier No. 2082131KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 115% H3PO4 basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8017-16-1
Molecular Formula (HO3P)n
MDL Number MFCD00084480
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name Polyphosphoric acid
SMILES [H]OP(O)(O)=O

Specifications

Color Colorless
Boiling Point 300°C
Density 2.06g/mL at 25°C
Packaging Poly Bottle
Quantity 1 kg
Linear Formula Hn+2PnO3n+1
Physical Form Liquid
UN Number UN3264
Percent Purity 115% (H3PO4 basis)
Grade Reagent
Chemical Name or Material Polyphosphoric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.