missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Keck™ POM Conical Joint Clips

Product Code. 10204241
Click to view available options
Color:
Blue
Green
Red
Turquoise
Yellow
Unit Size:
Pack of 10

Product Code. 10204241

Brand: Lenz Laborglasinstrumente™ 01103729

Only null left
Add to basket
Add to basket

Ensures a secure connection while preventing damage to glass tubing and increasing safety in the laboratory. Lenz Laborglasinstrumente™ Keck™ POM Conical Joint Clips are manufactured from temperature resistant polyoxymethylene (POM).

  • Good spring properties
  • Color coded by size
  • Will not corrode or bend out of shape

Specifications

Quantity 10/Pk.
Type Joint Clip
Material POM
For Use With (Equipment) For conical ground joints
Color Red
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.