missing translation for 'onlineSavingsMsg'
Learn More

Polymyxin B Sulfate, MedChemExpress

Product Code. p-200096708
Encompass_Preferred
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
This item is not returnable. View return policy

Product Code. 16451986

Brand: Medchem Express HYA0248

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Polymyxin B Sulfate is a cationic surfactant antibiotic agent.

Polymyxin B Sulfate is a cationic surfactant antibiotic agent.

Specifications

Chemical Name or Material Polymyxin B Sulfate
Physical Form Powder
CAS 1405-20-5
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C55H96N16O13.H2SO4 (Polymyxin B2 Sulfate)
Solubility Information H2O : ≥ 25 mg/mL
SMILES O=S(O)(O)=O.O=C(N[C@H](C(N[C@@](C(N[C@H](C(N[C@@]1([H])[C@H](O)C)=O)CCN)=O)([H])CCN)=O)CC(C)C)[C@H](NC([C@@H](NC([C@@](CCNC1=O)([H])NC([C@H](CCN)NC([C@H]([C@H](O)C)NC([C@H](CCN)NC(CCCCC(C)C)=O)=O)=O)=O)=O)CCN)=O)CC2=CC=CC=C2.O=C(N[C@H](C(N[C@@](C(N[C@H](C(N[C@@]3([H])[C@H](O)C)=O)CCN)=O)([H])CCN)=O)CC(C)C)[C@H](NC([C@@H](NC([C@@](CCNC3=O)([H])NC([C@H](CCN)NC([C@H]([C@H](O)C)NC([C@H](CCN)NC(CCCC[C@@H](C)CC)=O)=O)=O)=O)=O)CCN)=O)CC4=CC=CC=C4
Grade Research

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.