missing translation for 'onlineSavingsMsg'
Learn More

Polymyxin B Sulfate, MedChemExpress

Product Code. 16451986
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451986 500 mg 500mg
16412029 1 g 1g
16450748 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451986 Supplier Medchem Express Supplier No. HYA0248
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Polymyxin B Sulfate is a cationic surfactant antibiotic agent.

Polymyxin B Sulfate is a cationic surfactant antibiotic agent.

Specifications

Chemical Name or Material Polymyxin B Sulfate
Physical Form Powder
CAS 1405-20-5
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C55H96N16O13.H2SO4 (Polymyxin B2 Sulfate)
Solubility Information H2O : ≥ 25 mg/mL
SMILES O=S(O)(O)=O.O=C(N[C@H](C(N[C@@](C(N[C@H](C(N[C@@]1([H])[C@H](O)C)=O)CCN)=O)([H])CCN)=O)CC(C)C)[C@H](NC([C@@H](NC([C@@](CCNC1=O)([H])NC([C@H](CCN)NC([C@H]([C@H](O)C)NC([C@H](CCN)NC(CCCCC(C)C)=O)=O)=O)=O)=O)CCN)=O)CC2=CC=CC=C2.O=C(N[C@H](C(N[C@@](C(N[C@H](C(N[C@@]3([H])[C@H](O)C)=O)CCN)=O)([H])CCN)=O)CC(C)C)[C@H](NC([C@@H](NC([C@@](CCNC3=O)([H])NC([C@H](CCN)NC([C@H]([C@H](O)C)NC([C@H](CCN)NC(CCCC[C@@H](C)CC)=O)=O)=O)=O)=O)CCN)=O)CC4=CC=CC=C4
Grade Research

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.