missing translation for 'onlineSavingsMsg'
Learn More

Polymyxin B Antibody (45), Novus Biologicals™

Product Code. 18398662 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398662 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398662 Supplier Novus Biologicals Supplier No. NBP311216

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Polymyxin B Monoclonal specifically detects Polymyxin B in Human samples. It is validated for Western Blot, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Polymyxin B
Applications Western Blot, Immunoassay
Classification Monoclonal
Clone 45
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunoassay
Formulation Culture Medium
Gene Alias Aerosporin, Poly-Rx
Host Species Mouse
Immunogen The monoclonal antibody 45 reacts with Polymyxin B.
Purification Method Unpurified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgM
Show More Show Less
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.