missing translation for 'onlineSavingsMsg'
Learn More

Polymixin B Sulfate, Fisher BioReagents

Product Code. 10207607
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10207607 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10207607 Supplier Fisher BioReagents Supplier No. BP29541
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Interferes with cytoplasmic membrane

  • Polymixin B Sulfate is an antibiotic used to prevent bacterial contamination of cell cultures
  • Disrupts the permeability of the cytoplasmic membrane  which leads to cell death
  • This antibiotic is highly effective against gram-negative bacteria
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Specifications

Type Polymixin B Sulfate
Physical Form Solid
CAS 1405-20-5
Source Bacillus polymyxa
Loss on Drying < 7.0%
Packaging Amber Glass
Concentration >6000 U/mg
Quantity 1 g
Molecular Formula C48H84N16O17S
MDL Number MFCD00131991
Merck Index 15, 7693
InChI Key HNDFYNOVSOOGDU-UHFFFAOYNA-N
SMILES OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O
IUPAC Name 4-amino-N-{1-[(3-amino-1-{[6,9,18-tris(2-aminoethyl)-15-benzyl-3-(2-hydroxyethyl)-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl]carbamoyl}propyl)carbamoyl]-3-hydroxypropyl}-2-formamidobutanamide; sulfuric acid
Molecular Weight (g/mol) 1189.36
PubChem CID 133109994
Formula Weight 1385.61
Percent Purity >6000 U/mg
Chemical Name or Material Polymixin B Sulfate
Show More Show Less
compliance-icons
Product Identifier
  • Polymixin B sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.