missing translation for 'onlineSavingsMsg'
Learn More

Melford Polymixin B Sulfate

Product Code. 30018704
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018704 5 g 5g
30018703 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018704 Supplier Melford Supplier No. P401605.0
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Polymyxin B Sulfate is an antibiotic that belongs to the polymyxin class of antibiotics and is primarily effective against Gram-negative bacteria.

Polymyxin B Sulfate is an antibiotic that belongs to the polymyxin class of antibiotics and is primarily effective against Gram-negative bacteria.

Polymyxin B is derived from Bacillus polymyxa and acts by disrupting the integrity of the bacterial cell membrane.

In the field of microbiology and cell culture, Polymyxin B Sulfate is commonly used to prevent bacterial contamination. It is particularly effective against Gram-negative bacteria, including multidrug-resistant strains such as Pseudomonas aeruginosa. Researchers add Polymyxin B to growth media or solutions to inhibit the growth of these bacteria and maintain the purity of cultures.

Overall, Polymyxin B Sulfate is a valuable antibiotic in the life science marketplace, primarily used in microbiology, cell culture, and molecular biology research. Its effectiveness against Gram-negative bacteria and multidrug-resistant strains makes it a crucial tool for maintaining sterile cultures and studying bacterial pathogens. However, careful consideration of its potential toxicity is necessary when using it in cell culture applications.

Specifications

Chemical Name or Material Polymixin B Sulfate
CAS 1405-20-5
Quantity 5 g
Molecular Formula C55H96N16O13-2H2SO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.