missing translation for 'onlineSavingsMsg'
Learn More

Polymethylhydrosiloxane

Product Code. 10179552 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10179552 500 g Glass bottle 500g
10774823 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10179552 Supplier Thermo Scientific Acros Supplier No. 175095000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9004-73-3 | (CH4OSi)n | 60.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-73-3
Molecular Formula (CH4OSi)n
Molecular Weight (g/mol) 60.13
MDL Number MFCD00084478
InChI Key TXYXVOOPRILIHR-UHFFFAOYSA-N
Synonym polymethyl hydrosiloxane, 2-methyl-1,2-oxasilinanato, ch3 3sio ch3 hsio nsi ch3 3
PubChem CID 12487658
IUPAC Name 2-methyl-1,2$l^{3}-oxasilinane
SMILES C[SiH](-*)O-*

Specifications

Color Colorless
Physical Form Viscous Liquid
Linear Formula (CH3)3SiO[(CH3)HSiO]nSi(CH3)3
Density 1.0000g/mL
Solubility Information Solubility in water: practically insoluble. Other solubilities: may react, soluble in most common organic solvents, (should not be used in halocarbon solvent because, of the possibility of hydride-halide interchange, on catalysis or with heating)
Quantity 500 g
Flash Point 121°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3970 to 1.4010
Specific Gravity 1
Viscosity 15-30 mm2/s (25°C)
Chemical Name or Material Polymethylhydrosiloxane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.