missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygosil™ Phase

Brand:  Macherey-Nagel™ 711020.100

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10699561

  • £271.00 / 100g
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

10 μm
Liquid Chromatography
Polygosil Phases
60 Å
100 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers