missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygoprep™ 60 to 20 CN

Brand:  Macherey-Nagel™ 711016.100

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10698791

  • £170.00 / 100g
Estimated Shipment: 30-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

15 to 25 μm
POLYGOPREP 60 to 20
2 to 8
POLYGOPREP CN
Polygoprep
Liquid Chromatography
60 Å
Liquid Chromatography
0.1 kg
350 m2/g
Silica
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers