missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Polygoprep™ 100 to 30
£177.00
Specifications
Particle Size | 25 to 40 μm |
---|---|
Pore Size | 100 Å |
Stationary Phase | POLYGOPREP 100 to 30 |
Technique | Liquid Chromatography |
pH | 2 to 8 |
Specifications
25 to 40 μm | |
POLYGOPREP 100 to 30 | |
2 to 8 | |
280 m2/g | |
Silica |
100 Å | |
Liquid Chromatography | |
POLYGOPREP | |
Polygoprep | |
Liquid Chromatography |