missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygoprep™ 100 to 30 C18

Brand:  Macherey-Nagel™ 711032.1000

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10718232

  • £1200.00 / 1000g
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

25 to 40 μm
POLYGOPREP 100 to 30
2 to 8
POLYGOPREP C18
Polygoprep
Liquid Chromatography
100 Å
Liquid Chromatography
1 kg
280 m2/g
Silica
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers