missing translation for 'onlineSavingsMsg'
Learn More

Polygalacturonic acid, M.W. 25,000-50,000

Product Code. 11396836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396836 100 g 100g
11386836 25 g 25g
11306846 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396836 Supplier Thermo Scientific Alfa Aesar Supplier No. L04206.22
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25990-10-7 | (C6H8O6)n | 194.14 g/mol

Polygalacturonic acid is reagent which sequesters metal ions and produces gels that can be used for the production of biomolecules. It is used for drug release or to immobilize cells and enzymes . Polygalacturonic acid may be used to identify, differentiate and characterized plant cell wall degrading polygalacturonase(s) from fungi.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25990-10-7
Molecular Formula (C6H8O6)n
Molecular Weight (g/mol) 194.14
MDL Number MFCD00131972
InChI Key AEMOLEFTQBMNLQ-BKBMJHBISA-N
Synonym galacturonan, alpha-d-galactopyranuronic acid, calcium pectate, sodium pectate, alpha-d-galacturonic acid, calcium polygalacturonate, pectate, d-galacturonan, sulfated polygalacturonic acid, galacturonate
PubChem CID 445929
ChEBI CHEBI:33885
IUPAC Name (2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid
SMILES O[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O

Specifications

Quantity 100 g
Optical Rotation +250° (c=1 in 0.1N NaOH)
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 25 to 50 kd
Chemical Name or Material Polygalacturonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.