missing translation for 'onlineSavingsMsg'
Learn More

Polygalacturonic acid, >86%, MP Biomedicals™

Product Code. 11428370 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 11428370

Brand: MP Biomedicals 0210271190

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 25990-10-7
Molecular Formula (C6H8O6)n
Molecular Weight (g/mol) 194.14
MDL Number MFCD00131972
InChI Key AEMOLEFTQBMNLQ-BKBMJHBISA-N
Synonym galacturonan, alpha-d-galactopyranuronic acid, calcium pectate, sodium pectate, alpha-d-galacturonic acid, calcium polygalacturonate, pectate, d-galacturonan, sulfated polygalacturonic acid, galacturonate
PubChem CID 445929
ChEBI CHEBI:33885
IUPAC Name (2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid
SMILES O[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O

Specifications

Color Tan to White
Assay Percent Range >86%
Quantity 500 g
Formula Weight (176)n n=150 to 400
Percent Purity 88.7%
Physical Form Crystalline Powder
Chemical Name or Material Polygalacturonic acid, from Pectin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.