Learn More
Polyethyleneimine, branched, M.W. 70,000, 30% w/v aq. soln., Thermo Scientific Chemicals
A protein precipitant used to purify proteins
£57.40 - £1088.00
Chemical Identifiers
CAS | 9002-98-6 |
---|---|
Molecular Formula | ((C2H5N)zC2H4N)y(C2H5N)x |
Molecular Weight (g/mol) | 43.07 |
MDL Number | MFCD00084427 |
InChI Key | NOWKCMXCCJGMRR-UHFFFAOYSA-N |
Synonym | ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin |
PubChem CID | 9033 |
ChEBI | CHEBI:30969 |
IUPAC Name | aziridine |
SMILES | *-CCNCCN(-*)CCN-* |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11368528
|
Thermo Scientific Alfa Aesar
040529.14 |
25 g |
£57.40
25g |
In Stock
Log in to see stock. |
|||||
11358528
|
Thermo Scientific Alfa Aesar
040529.22 |
100 g |
£208.00
100g |
In Stock
Log in to see stock. |
|||||
11378528
|
Thermo Scientific Alfa Aesar
040529.36 |
500 g |
£1088.00
500g |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
Description
Acts as a protein precipitant used to purify proteins. It is used as chelating agent and as a scavenger for aldehydes and oxides. Also used in detergents, paper industry, dyes, printing inks and in water treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9002-98-6 | |
43.07 | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
9033 | |
aziridine |
((C2H5N)zC2H4N)y(C2H5N)x | |
MFCD00084427 | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
CHEBI:30969 | |
*-CCNCCN(-*)CCN-* |
Specifications
Colorless to Yellow | |
Liquid | |
MFCD00084427 | |
Soluble in hot water at a low pH,in methanol,ethanol,or chloroform,but insoluble in cold water,benzene,ethyl ether,and acetone. | |
*-CCNCCN(-*)CCN-* | |
43.07 | |
CHEBI:30969 | |
Odorless |
9002-98-6 | |
((C2H5N)zC2H4N)y(C2H5N)x | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
aziridine | |
9033 | |
30% w/v aq. soln. | |
Polyethyleneimine, branched, M.W. 10,000 |
Safety and Handling
missing translation for 'rtecsNumber' : TQ7915000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only