missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, branched, M.W. 600, 99%, Thermo Scientific Chemicals

Catalog Number 11308528
Click to view available options
Quantity:
25 g
100 g
500 g
9002-98-6
((C2H5N)zC2H4N)y(C2H5N)x
43.07
MFCD00084427
NOWKCMXCCJGMRR-UHFFFAOYSA-N
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
9033
CHEBI:30969
aziridine
*-CCNCCN(-*)CCN-*
9002-98-6
((C2H5N)zC2H4N)y(C2H5N)x
43.07
MFCD00084427
NOWKCMXCCJGMRR-UHFFFAOYSA-N
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
9033
CHEBI:30969
aziridine
*-CCNCCN(-*)CCN-*

Polyethyleneimine, branched, M.W. 600 acts as an auxiliary agent. It is used in paper-making, water treatment, detergents, adhesives and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Physical Form Liquid
Solubility Information Soluble in toluene,methanol,ethanol,chloroform and xylene. Insoluble in water,alcohol,benzene,ethyl ether and acetone.
Quantity 25 g
Percent Purity 99%
Odor Amine-like
Chemical Name or Material Polyethyleneimine, branched, M.W. 600

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'