Learn More
Polyethyleneimine, branched, M.W. 600, 99%
Description
Polyethyleneimine, branched, M.W. 600 acts as an auxiliary agent. It is used in paper-making, water treatment, detergents, adhesives and cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Physical Form | Liquid |
| Solubility Information | Soluble in toluene,methanol,ethanol,chloroform and xylene. Insoluble in water,alcohol,benzene,ethyl ether and acetone. |
| Quantity | 100 g |
| Percent Purity | 99% |
| Odor | Amine-like |
| Chemical Name or Material | Polyethyleneimine, branched, M.W. 600 |
Safety and Handling
- Polyethyleneimine, branched, M. W. 600
- LONG-TERM AQUATIC HAZARD Chronic 2
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Polyethyleneimine
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.