missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, branched, M.W. 2,000

Product Code. 11328528
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328528 500 g 500g
11348528 25 g 25g
11338528 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11328528 Supplier Thermo Scientific Alfa Aesar Supplier No. 040528.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol

Polyethyleneimine is used for various applications in products like detergents, adhesives, water treatment agents and cosmetics. It is also used as flocculating agent, enzyme carrier, wet-strength agent in the paper-making process, chelating agent form complexes with metal ions such as zinc and zirconium. It acts as scavenger for aldehydes and oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*

Specifications

Color Colorless to Yellow
Physical Form Liquid
Solubility Information Soluble in hot water at a low pH,in methanol,ethanol,or chloroform,but insoluble in cold water,benzene,ethyl ether,and acetone.
Quantity 500 g
Percent Purity 99%
Odor Amine-like
Chemical Name or Material Polyethyleneimine, branched, M.W. 1,800

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.