Learn More
Masterflex™ Polyethylene Straight Connector, pack of 100
Product Code. p-7116387
Click to view available options
Tubing Size:
3/16 in.
5/16 in.
5/32 in.
Unit Size:
Pack of 100
Product Code. 11709669
Brand: Masterflex™ 3062587
Only null left
Add to basket
Add to basket
Designed for quick and easy assembly without tools
  • Adapts from a NPT male thread to a hose barb
  • Barbed fittings are designed for quick and easy assembly without tools
  • Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
  • Available in packs of ten
Specifications
Description Barb Connection
Autoclavable Not Autoclavable
Material HDPE
Tubing Size 5/16 in.
Hose Barb 5/16 in.
Port Size 5/16 in.
Sterilization Method Ethylene Oxide
Color Translucent
Product Type Straight Connector