Learn More
Kartell™ Polyethylene Short Spout Dropping Bottle Screw Caps
Product Code. 10132571
Change view
Click to view available options
Thread Size (Metric):
GL 32
Unit Size:
Pack of 50
Product Code. Thread Size (Metric) unitSize
10132571 GL 32 Pack of 50
1 options
Product Code. 10132571 Supplier Kartell™ Supplier No. 0330500
Manufactured from polyethylene for strength and fit. Kartell™ Short Spout Dropping Bottle Screw Caps provide a sturdy solution for laboratory needs and are available in a variety of sizes.
  • Fit narrow neck standardized bottles Kartell™ 15801, 15821, 15841, 15861, 15881, 15901
  • Supplied with PE cap for tip closure
  • Spout length of 19mm
Specifications
Includes PE cap for tip closure
Material Polyethylene
Screw Material Polyethylene
Product Line Cap
Thread Size (Metric) GL 32
Neck Finish 30 mm
Product Type Short Spout Dropping Bottle Screw Cap
For Use With (Equipment) Narrow neck and wide neck bottle