Learn More
Kartell™ Polyethylene Dropping Bottle Screw Caps
Product Code. 10091021
Change view
Click to view available options
Thread Size (Metric):
GL 18
GL 25
Length (Metric):
16mm
40mm
41mm
Product Type:
Dropping Bottle Screw Cap
Screw Cap
Unit Size:
Each
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Length (Metric) Product Type unitSize
10091021 GL 18 16mm Dropping Bottle Screw Cap Pack of 100
11372914 GL 18 40mm Screw Cap Each
11382914 GL 25 41mm Dropping Bottle Screw Cap Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10091021 Supplier Kartell™ Supplier No. 0330300
Only null left
Add to basket
Add to basket
Manufactured from polyethylene for strength and fit. Kartell™ Polyethylene Dropping Bottle Screw Caps, available in a variety of sizes and spout styles, provide a sturdy solution for laboratory dropping needs.
  •  Available in long or short dropping spout
  • Fits standardized narrow and wide neck bottles
  • Precision dropping for exact measurements with long spout
  • Steady stream for filling small quantities with short spout
  • Supplied with PE tip closure cap to avoid spillage
Specifications
Includes PE cap for tip closure
Material Polyethylene
Length (Metric) 16mm
Quantity 100Pack
Thread Size (Metric) GL 18
Neck Finish 30 mm
Product Type Dropping Bottle Screw Cap
For Use With (Equipment) Narrow neck standardized bottles part. no. 15801 - 15821 - 15841 - 15861 - 15881 - 15901 and wide neck standardized bottles part. no. 1608 - 1610
Closure Type Short Spout