Learn More
Hirschmann™ Polyethylene Poly Stopper
Product Code. 30090415
Change view
Click to view available options
Standard Taper Joint:
NS 10/19
NS 12/21
NS 14/23
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
30090415 NS 14/23 Pack of 10
30090421 NS 7/16 Pack of 10
30090401 NS 10/19 Pack of 10
30090407 NS 12/21 Pack of 10
10640992 NS 45/40 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30090415 Supplier Hirschmann™ Supplier No. 8900270
  • ONLINE EXCLUSIVE PRICE
    £18.02
    £15.55 / Pack of 10
    Save £2.47
    13% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
  • PE
  • Available to fit a range of sizes
Specifications
Quantity 1 Ea.
Standard Taper Joint NS 14/23
Material Polyethylene
Product Type Polyethylene Stopper
Shape Hexagonal Grip
Sterility Non-sterile