missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene oxide, M.W. 1,000,000, Thermo Scientific Chemicals

Product Code. 11305917
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Polyethylene oxide is used as lubricants for rubber molds, textiles, metalworking and ceramics production. Further, it is used in food packaging, water-based paints, polishes, paper coatings, dispersing agents and chemical intermediates. In addition, it is used to impart viscosity to and modify the flow of aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Weight (g/mol) 62.07
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 5 to 7
Physical Form Powder
Molecular Formula (C2H4O)n
MDL Number MFCD01779601
Merck Index 14,7568
Solubility Information Soluble in organic solvents,aromatic hydrocarbons and water. Slightly soluble in aliphatic hydrocarbons.
Quantity 25 g
Odor Odorless
Chemical Name or Material Polyethylene oxide, M.W. 1,000,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'