missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene oxide, M.W. 1,000,000, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Polyethylene oxide is used as lubricants for rubber molds, textiles, metalworking and ceramics production. Further, it is used in food packaging, water-based paints, polishes, paper coatings, dispersing agents and chemical intermediates. In addition, it is used to impart viscosity to and modify the flow of aqueous solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
pH | 5 to 7 |
Physical Form | Powder |
Molecular Formula | (C2H4O)n |
MDL Number | MFCD01779601 |
Merck Index | 14,7568 |
Solubility Information | Soluble in organic solvents,aromatic hydrocarbons and water. Slightly soluble in aliphatic hydrocarbons. |
Quantity | 100 g |
Odor | Odorless |
Chemical Name or Material | Polyethylene oxide, M.W. 1,000,000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'