Learn More
Pal International™ Deluxe Smooth Sole Fabric Overshoes
Product Code. 12963071
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 200
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12963071 White Pack of 250
12943071 White Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12963071 Supplier Pal International™ Supplier No. N68110CN
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for a better fit and improved comfort. Pal International™ Deluxe Smooth Sole Fabric Overshoes are especially suitable for labarotories or for visitors to hygienic manufacturing sites.
  • Manufactured from strong and durable non-woven polypropylene.
  • Higher spec overshoe designed for a better fit and improved comfort.
  • Especially suitable for more sensitive areas such as labarotories or for visitors to hygienic manufacturing sites.
  • Embossed sole gives extra grip and durability.
  • Elasticized band for easy wear and removal
  • DIsposable
  • One size fits all.
  • Package of 200
Specifications
Color White
Heel Style Elasticated ankle
Material Polypropylene
Size Universal
Sole Features Smooth
Product Type Overshoe