missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polyethylene N 18 Snap Cap

Product Code. 10136831 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Transparent
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10136831 Transparent Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10136831 Supplier Macherey-Nagel™ Supplier No. 70274
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Transparent
Material Polyethylene
For Use With (Application) For use with 70271 and 70272
Insert Type Snap Cap
Quantity 100 Pk.
Closure Size N18
Product Type Snap Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.