missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Trimethylnonyl Ether (Technical Grade), TRC

Product Code. 30455640
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455640 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455640 Supplier Toronto Research Chemicals Supplier No. TRCT54881310ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polyethylene Glycol Trimethylnonyl Ether (Technical Grade)
CAS 68131-40-8
Quantity 10 mL
Synonym Alcohols','C11-15-Secondary','Ethoxylated, Ethoxylated C11-15-Secondary AlcoholsEthoxylated C11-15-Secondary Alcs.
SMILES [2H]C([2H])([2H])CCCCc1cc(O)c2[C@@H]3C=C(C)CC[C@H]3C(C)(C)Oc2c1
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.