missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol, MW 8000, , Molecular biology reagent grade, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
2 kg
Unit Size:
2kg
Specifications
Specifications
| Color | White |
| CAS | 25322-68-3 |
| Physical Form | Prills |
| Molecular Formula | (C2H4O)n |
| MDL Number | MFCD01779601 |
| Synonym | Carbowax, Alkapol PEG-8000 |
| Quantity | 2 kg |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| SMILES | [H]OCCO |
| IUPAC Name | ethane-1,2-diol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction