missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol, MP Biomedicals™

Product Code. 11466591 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466591 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11466591 Supplier MP Biomedicals Supplier No. 02194839.5
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Also known as PEG. Polyethylene Glycol, MP Biomedicals is a condensation polymer of ethylene oxide and water. It is used in precipitation of proteins, separating and purifying biomolecules, induction of cell hybridization, and as a fusing agent in enhanci

PEGs are susceptible to oxidative degradation in the presence of air. Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen, or addition of an antioxidant can limit the amount of degradation. PEGs do not hydrolyze or deteriorate upon storage. PEGs do not support the growth of molds.

Specifications

CAS 25322-68-3
Physical Form White Prills
Molecular Formula (C2H4O)n
MDL Number MFCD01779601
Synonym Carbowax, PEG, Poly(Ethylene Oxide), Alkapol PEG-8000, Macrogol, α-Hydro-o-hydroxypoly (oxy-1,2-ethanediyl)
Solubility Information Soluble in water (approximately 630mg/mL, 20°C). PEGs are also soluble in many polar solvents such as acetone, alcohols and chlorinated solvents. They are insoluble in nonpolar solvents such as hydrocarbons.
Quantity 500 g
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES [H]OCCO
IUPAC Name ethane-1,2-diol
Molecular Weight (g/mol) 62.07
Formula Weight 8000g/mol
Grade Molecular Biology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.