missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol, MP Biomedicals™

Product Code. 11425372 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425372 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11425372 Supplier MP Biomedicals Supplier No. 02195445.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be used to precipitate DNA. Polyethylene Glycol, MP Biomedicals (PEG) is a condensation polymer of ethylene oxide and water. PEGs are susceptible to oxidative degradation in the presence of air. PEGs do not hydrolyze or deteriorate upon storage. PEGs

PEG has been used in the precipitation of proteins, as a fusing agent in enhancing the effect of macrophages on hybridoma, in the separation and purification of biomolecules, and in induction of cell hybridization. PEG 8000 can be used to precipitate DNA. PEG has also been used to recover pathogenic viruses and human rotaviruses from water, oyster and sediment samples. PEG has demonstrated to have anti-cell proliferation and antitumor effects via induction of apoptosis.

  • PEGs are susceptible to oxidative degradation in the presence of air
    • Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen, or addition of an antioxidant, can limit the amount of degradation
  • PEGs do not hydrolyze or deteriorate upon storage
  • PEGs do not support the growth of molds
  • PEG is incompatible with phenol and may reduce the antimicrobial action of other preservatives
TRUSTED_SUSTAINABILITY

Specifications

CAS 25322-68-3
Melting Point 56°C to 66°C
Density 1.1-1.2g/mL (Lit.)
pH 5.0 to 7.0 (5% aq. soln.)
Molecular Formula (C2H4O)n
MDL Number MFCD01779601
Quantity 500 g
Synonym Carbowax, PEG, Poly(Ethylene Oxide), Alkapol PEG-200
Solubility Information Soluble in aqueous solution (50mg/mL, clear, colorless solution).
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES [H]OCCO
IUPAC Name ethane-1,2-diol
Molecular Weight (g/mol) 62.07
Formula Weight ∼8000
Vapor Pressure <0.01mmHg at 20°C (Lit.)
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.