missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol monomethylether, 1,900

Product Code. 11388678
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388678 100 g 100g
11378678 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388678 Supplier Thermo Scientific Alfa Aesar Supplier No. 041563.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol

Polyethylene glycol monomethylether is used in the introduction of hydrophilic sites into polymers. It is also used to stabilize polymer emulsions. Further, it is employed in the synthesis of comb polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-74-4
Molecular Formula (C2H4O)nCH4O
Molecular Weight (g/mol) 76.10
MDL Number MFCD00084416
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Color White to Yellow
Physical Form Solid Mass
Melting Point 52°C
Solubility Information Slightly miscible with water.
Quantity 100 g
Flash Point >110°C (230°F)
Chemical Name or Material Polyethylene glycol monomethylether 1,900

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.