missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol monomethyl ether methanesulfonate, M.W. 1,000, Thermo Scientific Chemicals
POA - £246.00
Chemical Identifiers
CAS | 175172-61-9 |
---|---|
Molecular Formula | C4H10O4S |
Molecular Weight (g/mol) | 154.18 |
MDL Number | MFCD03100948 |
InChI Key | BCKAHDGFNHDQST-UHFFFAOYSA-N |
PubChem CID | 146293 |
IUPAC Name | 2-methoxyethyl methanesulfonate |
SMILES | COCCOS(=O)(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15427215
|
Thermo Scientific Alfa Aesar
046437.MD |
250 mg | |||||||
15407215
|
Thermo Scientific Alfa Aesar
046437.03 |
1 g |
£54.30
1g |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15417215
|
Thermo Scientific Alfa Aesar
046437.06 |
5 g |
£246.00
5g |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
175172-61-9 | |
154.18 | |
BCKAHDGFNHDQST-UHFFFAOYSA-N | |
2-methoxyethyl methanesulfonate |
C4H10O4S | |
MFCD03100948 | |
146293 | |
COCCOS(=O)(=O)C |
Specifications
White | |
Granular Solid | |
MFCD03100948 | |
BCKAHDGFNHDQST-UHFFFAOYSA-N | |
2-methoxyethyl methanesulfonate | |
146293 |
175172-61-9 | |
C4H10O4S | |
methanesulfonic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, methanesulfonate, 2-methoxyethanol methanesulfonate, 2-methoxyethyl mesylate, 2-methoxyethylmethanesulfonate, acmc-1c990, 2-methoxyethyl methane sulfonate, methanesulfonic acid2-methoxyethyl ester, ethanol, 2-methoxy-, 1-methanesulfonate, methanesulfonic acid 2-methoxy-ethyl ester | |
COCCOS(=O)(=O)C | |
154.18 | |
Polyethylene glycol monomethyl ether methanesulfonate, M.W. 1,000 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only