missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Monoisotridecyl Ether, TRC

Product Code. 30412606
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412606 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412606 Supplier Toronto Research Chemicals Supplier No. TRCP6882801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polyethylene Glycol Monoisotridecyl Ether
CAS 9043-30-5
Molecular Formula C13H28O•(C2H4O)n
Quantity 1 g
Synonym α-Isotridecyl-ω-hydroxypoly(oxy-1,2-ethanediyl), Arlypon IT 10, Berol 048, Dehscoxid 732, Ethox 2400, Ethox TDA 9, Ethoxylated isotridecanol, Ethoxylated Isotridecyl Alcohol, Eusapon S, Exxal F 5716, Genapol V 4739, Genapol X, Gezetol 138, Imbentin T 050, Isotridecanol Ethoxylate, Isotrideceth 15, Leocol TD 120, Leocol TD 150, Leocol TD 50, Leocol TD 90, Leocol TDA 400-75, Lutensol TO 10, M 0-7, M 0-9, Marlipal 013/40, Marlosol TA 3050, Marlosol TA 3090, Nissan Dispanol TOC, Noigen TDS 200D, Noigen TDS 80, Novanit MA, Novel II TDA 8.5, Novel II TDA 9, Novel II TDA 9.5, PEG isotridecyl ether, Polyethylene glycol isotridecyl ether, Polyethylene Glycol Isotridecyl Monoether, Polyethylene Glycol Monoether with isotridecyl alcohol, Polyethylene Glycol Monoisotridecyl ether, Polyoxyethylene isotridecyl ether, Rhodasurf ROX, Rhodasurf TR 15-40, Rhodasurf TR 25-25, Rokanol IT, Surfonic TDA 11, Surfonic TDA 15/88, Surfonic TDA 3, Surfonic TDA 8, Surfonic TDA 9, Synperonic 13/10, Synperonic 13/6.5, T-DET A 134, TA 400, Tego Alkanol TD 6, Unitol IT 140, Villa 51, Walloxen TR 100, Walloxen TR 16, X 1005
InChI Formula InChI=1S/C30H62O10/c1-2-3-4-5-6-7-8-9-10-11-13-32-15-17-34-19-21-36-23-25-38-27-29-40-30-28-39-26-24-37-22-20-35-18-16-33-14-12-31/h31H,2-30H2,1H3
SMILES OCCO[(C13H27-iso)]
Molecular Weight (g/mol) 582.8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.