missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol methyl ether methacrylate, M.W. 5,000

Product Code. 15437335
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437335 5 g 5g
15447335 250 mg 250mg
15427335 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15437335 Supplier Thermo Scientific Alfa Aesar Supplier No. 046838.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26915-72-0 | C13H24O6 | 276.329 g/mol

Polyethylene glycol methyl ether methacrylate, is used as Emulsion polymers, cosmetics, Construction, emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26915-72-0
Molecular Formula C13H24O6
Molecular Weight (g/mol) 276.329
MDL Number MFCD00241432
InChI Key KRCGBOKYIUDIFY-UHFFFAOYSA-N
Synonym 2-methoxyethyl methacrylate, beta-methoxyethyl methacrylate, methacrylic acid, 2-methoxyethyl ester, 2-propenoic acid, 2-methyl-, 2-methoxyethyl ester, poly ethylene glycol methyl ether methacrylate, pegmem, peg-mem, acmc-209oay, methoxyethyl methacrylate, 2-methoxyethylmethacrylate
PubChem CID 93675
IUPAC Name 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOCCOCCOCCOC

Specifications

Color White
Physical Form Granular Solid
Solubility Information It is soluble in water.
Quantity 5 g
Chemical Name or Material Polyethylene glycol methyl ether methacrylate, M.W. 5,000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.