missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol methyl ether acrylate, M.W. 5,000

Product Code. 15427195
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427195 5 g 5g
15437195 250 mg 250mg
15417195 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15427195 Supplier Thermo Scientific Alfa Aesar Supplier No. 046388.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32171-39-4 | (C2H4O)nC4H6O2 | NaN g/mol

Polyethylene glycol methyl ether acrylate is used as a raw material in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32171-39-4
Molecular Formula (C2H4O)nC4H6O2
Molecular Weight (g/mol) NaN
MDL Number MFCD00803685
InChI Key HFCUBKYHMMPGBY-UHFFFAOYSA-N
Synonym 2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
PubChem CID 18392
IUPAC Name 2-methoxyethyl prop-2-enoate
SMILES COCCOC(=O)C=C

Specifications

Color White
Physical Form Granular Solid
Solubility Information Soluble in water
Quantity 5 g
Chemical Name or Material Polyethylene glycol methyl ether acrylate, M.W. 5,000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.