missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol methyl ether acrylate, M.W. 1,000, Thermo Scientific Chemicals

Product Code. 15497245
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Polyethylene glycol methyl ether acrylate is used as a raw material in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32171-39-4
Molecular Formula (C2H4O)nC4H6O2
Molecular Weight (g/mol) NaN
MDL Number MFCD00803685
InChI Key HFCUBKYHMMPGBY-UHFFFAOYSA-N
Synonym 2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
PubChem CID 18392
IUPAC Name 2-methoxyethyl prop-2-enoate
SMILES COCCOC(=O)C=C

Specifications

Color White
Physical Form Granular Solid
Solubility Information Soluble in water
Quantity 250 mg
Chemical Name or Material Polyethylene glycol methyl ether acrylate, M.W. 1,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'