missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol Glycidyl Lauryl Ether, TRC
Specifications
Specifications
| Chemical Name or Material | Polyethylene Glycol Glycidyl Lauryl Ether |
| CAS | 86630-59-3 |
| Molecular Formula | (C2H4O)n . C15H30O2 |
| Quantity | 2.5 g |
| Synonym | Poly(oxy-1,2-ethanediyl)','α-dodecyl-ω-(oxiranylmethoxy)- (9CI), Denacol EX 171, Denacol FCA 050, Emulgen 120GE, EX 171, Lauryl (EO)15 glycidyl ether, Polyethylene glycol dodecyl glycidyl ether, Polyethylene glycol glycidyl lauryl ether, Polyethylene glycol lauryl glycidyl ether, Polyethylene glycol monododecyl monoglycidyl ether, Polyethylene glycol monoglycidyl monolauryl ether, Polyoxyethylene glycidyl lauryl ether |
| InChI Formula | InChI=1S/C17H34O3/c1-2-3-4-5-6-7-8-9-10-11-12-18-13-14-19-15-17-16-20-17/h17H,2-16H2,1H3 |
| SMILES | CCCCCCCCCCCCOCCOCC1CO1 |
| Molecular Weight (g/mol) | 286.4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?