missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol dimethanesulfonate, M.W. 1,000, Thermo Scientific™
£17.18 - £198.00
Chemical Identifiers
CAS | 52410-74-9 |
---|---|
Molecular Formula | C4H10O6S2 |
Molecular Weight (g/mol) | 218.238 |
MDL Number | MFCD03100947 |
InChI Key | QSQFARNGNIZGAW-UHFFFAOYSA-N |
Synonym | ethylene dimethanesulfonate, dmse, 1,2-bis mesyloxy ethane, ethylene methanesulfonate, ethylene glycol dimesylate, ethane dimethane sulphonate, 1,2-ethanediyl dimethanesulfonate, dimethylene dimethanesulfonate, ethylene bis methanesulfonate, ethane dimethanesulphonate |
PubChem CID | 20796 |
IUPAC Name | 2-methylsulfonyloxyethyl methanesulfonate |
SMILES | CS(=O)(=O)OCCOS(=O)(=O)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
52410-74-9 | |
218.238 | |
QSQFARNGNIZGAW-UHFFFAOYSA-N | |
20796 | |
CS(=O)(=O)OCCOS(=O)(=O)C |
C4H10O6S2 | |
MFCD03100947 | |
ethylene dimethanesulfonate, dmse, 1,2-bis mesyloxy ethane, ethylene methanesulfonate, ethylene glycol dimesylate, ethane dimethane sulphonate, 1,2-ethanediyl dimethanesulfonate, dimethylene dimethanesulfonate, ethylene bis methanesulfonate, ethane dimethanesulphonate | |
2-methylsulfonyloxyethyl methanesulfonate |
Specifications
White | |
Waxy Solid | |
MFCD03100947 | |
QSQFARNGNIZGAW-UHFFFAOYSA-N | |
2-methylsulfonyloxyethyl methanesulfonate | |
20796 |
52410-74-9 | |
C4H10O6S2 | |
ethylene dimethanesulfonate, dmse, 1,2-bis mesyloxy ethane, ethylene methanesulfonate, ethylene glycol dimesylate, ethane dimethane sulphonate, 1,2-ethanediyl dimethanesulfonate, dimethylene dimethanesulfonate, ethylene bis methanesulfonate, ethane dimethanesulphonate | |
CS(=O)(=O)OCCOS(=O)(=O)C | |
218.238 | |
Polyethylene glycol dimethanesulfonate, M.W. 1,000 |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures