missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol dimethacrylate, M.W. 3,400

Product Code. 15467295
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467295 250 mg 250mg
15422757 1 g 1g
15432757 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15467295 Supplier Thermo Scientific Alfa Aesar Supplier No. 046728.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25852-47-5 | CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3

Polyethylene glycol dimethacrylate is used as adhesives, coatings, sealants, photo resists, solder masks and photopolymers. Polyethylene Glycol Dimethacrylate forms homopolymers and copolymers. Copolymers of Polyethylene Glycol Dimethacrylate can be prepared with acrylic acid and its salts, amides and esters, and with methacrylates, acrylonitrile, maleic acid esters, vinyl acetate, vinyl chloride, vinylidene chloride, styrene, butadiene, unsaturated polyesters and drying oils. It is also used as a very useful feedstock for chemical syntheses. It is employed as a cross linker for PVC plastisols. Polyethylene Glycol 200 Dimethacrylate is incorporated in cosmetic encapsulation resins and dental polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25852-47-5
MDL Number MFCD00081877
Synonym ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate

Specifications

Color White
Physical Form Granular Solid
Molecular Formula CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3
Solubility Information It is soluble in water.
Quantity 250 mg
Sensitivity Light and moisture sensitive
Chemical Name or Material Polyethylene glycol dimethacrylate, M.W. 3,400

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.