missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 8000 (PEG), Fisher BioReagents™

Product Code. p-27885
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
100g
1kg
This item is not returnable. View return policy

Product Code. 10224963

Brand: Fisher BioReagents BP233100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Polyethylene Glycol 8000, HO-[CH2CH2O]n-H, CAS Number-25322-68-3, Polyethylene Oxide, Carbowax, PEG 8000, 1kg, Gray, White, 7000 to 9000, CAUTION!

PEG 8000 is suitable as a medium for the fusion of mammalian cells. It is also used to precipitate bacteriophage from lysed cell supernatants.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG 8000, Polyethylene Oxide, Carbowax™
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
pH 5.0 to 7.0
Physical Form Solid
Melting Point 55°C
Linear Formula H(OCH2CH2)nOH
Merck Index 15, 7688
Quantity 100 g
Formula Weight 7000 to 9000
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Polyethylene Glycol 8000
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.