missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol 8,000

Product Code. p-7058882
Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
Unit Size:
100g
10kg
2kg
500g
This item is not returnable. View return policy

Product Code. 11375747

Brand: Thermo Scientific Alfa Aesar 043443.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene glycol is used in the modification of therapeutic proteins and peptides to enhance its solubility. It is also used in the fabrication of bioactive and immuno isolating barriers for encapsulation of cells. In gas chromatography, it is used as a polar stationary phase. It is used as mold release agent and lubricant in fabricating elastomers. As a dispersant, it is used in toothpastes. It finds application in blood banking as a potentiator, which is used to detect antigens and antibodies. In pharmaceutical industry, it is used as tablet and capsule lubricant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG 8000
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
pH 5 to 7
Physical Form Powder
Melting Point 55°C to 63°C
Merck Index 14,7568
Solubility Information Soluble in water and aromatic hydrocarbons. Slightly soluble in aliphatic hydrocarbons and organic solvents.
Quantity 100 g
Odor Odorless
Chemical Name or Material Polyethylene glycol 8,000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.