missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol 6,000

Product Code. 11428177
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428177 1000 g 1000g
11438177 250 g 250g
11448177 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428177 Supplier Thermo Scientific Alfa Aesar Supplier No. A17541.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene glycol is used in the modification of therapeutic proteins and peptides to enhance its solubility. It is also used in the fabrication of bioactive and immuno isolating barriers for encapsulation of cells. In Gas chromatorgraphy, it is used as a polar stationary phase. It is used as mold release agent and lubricant in fabricating elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Physical Form Powder
Melting Point 60°C to 63°C
Merck Index 14,7568
Solubility Information Soluble in water and aromatic hydrocarbons. Slightly and aliphatic hydrocarbons.
Quantity 1000 g
Chemical Name or Material Polyethylene Glycol 6000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.