missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol 400

Product Code. 11439467
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439467 1000 g 1000g
11449467 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439467 Supplier Thermo Scientific Alfa Aesar Supplier No. B21992.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene glycol is used in a variety of pharmaceutical formulations. It finds application in Hewlett-Packard designjet printers as an ink solvent and a lubricant for the print heads. It is an important raw material for the manufacture of polyuretnae, PEG salts of lauric, oleic and stearic acids and latex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG 400
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 5 to 7
Melting Point 4°C to 8°C
Linear Formula H(OCH2CH2)nOH
Density 1.126
Merck Index 14,7568
Solubility Information Miscible with water,acetone,alcohols,benzene,glycerin,glycols and aromatic hydrocarbons. Slightly miscible with aliphatic hydrocarbons.
Quantity 1000 g
Flash Point 240°C (464°F)
Odor Odorless
Refractive Index 1.466
Sensitivity Hygroscopic
Chemical Name or Material Polyethylene glycol 400
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.