missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol 4,000, Thermo Scientific Chemicals

Catalog Number 11453207
Click to view available options
Quantity:
250 g
1000 g
5000 g
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG 4000
ethane-1,2-diol
[H]OCCO
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
PEG 4000
ethane-1,2-diol
[H]OCCO

Polyethylene glycol is used in gene delivery of protoplast transfection and incubation. It is used in the preparation of extraction for measuring cytosolic phosphoenolpyruvate carboxykinase (PEPCK) from plant tissues. It is useful to mediate DNA transformation into protoplasts derived from rice leaf sheaths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
pH 5 to 7
Melting Point 58°C to 62°C
Merck Index 14,7568
Solubility Information Soluble in water and organic solvents. Insoluble in aliphatic hydrocarbons.
Quantity 1000 g
Odor Odorless
Chemical Name or Material Polyethylene glycol 4,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'