missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 3350, Powder, BAKER™, J.T. Baker™

Product Code. 15588824 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
2 kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15588824 1 kg Wide Mouth Amber Glass Bottle Each
15598824 2 kg Poly Pail Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15588824 Supplier Avantor J.T.Baker Supplier No. U22108

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Physical Form Powder
Quantity 1 kg
Formula Weight 3000 to 3700 (Average)
Packaging Wide Mouth Amber Glass Bottle
Chemical Name or Material Polyethylene Glycol 3350
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.