missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 200 Grade, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7073399 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
25 L
Packaging:
Amber glass bottle
Plastic container
Unit Size:
25 L
500mL
This item is not returnable. View return policy

Product Code. 10487260

Brand: Fisher Chemical P/3675/08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 5.0 to 7.0
Physical Form Liquid
Melting Point -65°C
Quantity 500 mL
Formula Weight 200g/mol
Packaging Amber glass bottle
Vapor Pressure <0.01mmHg at 20°C
Viscosity 60 to 70 mPaS at 20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.